Recombinat Protein,Human IFN-alpha beta R2 Protein, Fc Tag,Human IFNAR2, Fc Tag (IF2-H5255) is expressed from human 293 cells (HEK2 93). It contains AA Ile 27 - Lys 243 (Accession NP997467).This protein carries a human IgG1 Fc tag at the C-terminus. 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human IFN-alpha beta R2 Protein, Fc Tag,Human IFNAR2, Fc Tag (IF2-H5255) is expressed from human 293 cells (HEK2 93). It contains AA Ile 27 - Lys 243 (Accession NP997467).This protein carries a human IgG1 Fc tag at the C-terminus. 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human IFN-alpha beta R2 Protein, Fc Tag,Human IFNAR2, Fc Tag (IF2-H5255) is expressed from human 293 cells (HEK2 93). It contains AA Ile 27 - Lys 243 (Accession NP997467).This protein carries a human IgG1 Fc tag at the C-terminus. 1 * 1 mg ACRBIF2-H5255-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou