Cynomolgus PCSK9, His Tag (PC9-C5223) is expressed from human 293 cells (HEK293). It contains AA Gln 31 - Gln 692 (Accession G7NVZ1 -1). 1 * 50 G
Fournisseur: ACROBIOSYSTEMS
Cynomolgus PCSK9, His Tag (PC9-C5223) is expressed from human 293 cells (HEK293). It contains AA Gln 31 - Gln 692 (Accession G7NVZ1 -1). 1 * 50 G
COMMANDE
Développer tout / Réduire tout
Cynomolgus PCSK9, His Tag (PC9-C5223) is expressed from human 293 cells (HEK293). It contains AA Gln 31 - Gln 692 (Accession G7NVZ1 -1). 1 * 50 µG ACRBPC9-C5223-50UG 1 * 50 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou