Mouse PD-L1, His Tag (PD1-M5220) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Thr 238 (Accession NP068693). 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Mouse PD-L1, His Tag (PD1-M5220) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Thr 238 (Accession NP068693). 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Mouse PD-L1, His Tag (PD1-M5220) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Thr 238 (Accession NP068693). 1 * 1 mg ACRBPD1-M5220-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou