GC columns, J&W VF-5ms
Fournisseur: Agilent
CP9043
HEWLCP9043EA 600 EUR InStock
HEWLCP9043
GC columns, J&W VF-5ms
Colonnes pour chromatographie
VF-5 ms is an inert and low bleed 5% phenyl-methyl phase for increased sensitivity, accuracy, and instrument uptime for semipolar and polar compounds.
  • 0.15 mm int.Ø columns available for high efficiency GC and GC/MS analysis
  • QC test results for retention index, efficiency, selectivity and bleed is reported with every column
The column has the lowest guaranteed bleed specification of 1 pA at 325 °C (30 m, 0.25 mm, 0.25 µm). VF-5ms has a slightly higher polarity than the VF-1 ms, resulting in a better selectivity for aromatic compounds. This selectivity, combined with superior inertness, also makes these columns applicable for a wide range of semipolar and even polar components, such as phenols.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité