J66886.06
J66886.06EA
172
EUR
InStock
J66886.06
Ceftiofur sodium 95%
Ceftiofur sodium
Purity: 95%
CAS No.: 104010-37-9
Molecular Formula: C19H16N5NaO7S3
Formula Weight: 545.54
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: Off-white or yellow
Solubility: Freely soluble in aqueous solution
MDL No.: MFCD01766184
Applications: A broad spectrum antibiotic