L19358.06
L19358.14
L19358.06EA
40.9
EUR
InStock
L19358.06
L19358.14
1-Formyl-4-methylpiperazine 98%
1-Formyl-4-methylpiperazine
Purity: 98%
CAS No.: 7556-55-0
Molecular Formula: C6H12N2O
Formula Weight: 128.17
Flash Point: 110°(230°F)
Refractive Index: 1.4910
Boiling Point: 236-238°
Beilstein Reference: 113020
MDL No.: MFCD00085930