06846-50MG
06846-50MGEA
50.9
EUR
InStock
06846-50MG
Supelco® TraceCERT® Organic Reference Standard, Chlorimuron Ethyl
Étalons
Étalons organiques
Chlorimuron-ethyl is a member of sulfonylurea herbicides used wildly for pre- and post-emergence control of broad-leaved weeds in soybeans maize peanuts beans and other crops. It is characterised by very low application rates high herbicidal activity good crop selectivity and low toxicity. Chlorimuron-ethyl is an acetolactate synthase (ALS) inhibitor that acts by inhibiting the biosynthesis of the essential amino acids like valine leucine and isoleucine. This stops the plant growth as cell division of growing roots and shoots tips are inhibited.
Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
Attention: P273 - P391 - P501