Nitrile gloves, Microflex® 93-850
Fournisseur: Ansell
Microflex®
112-5171EA 28.2 EUR InStock
112-5171 112-5167 112-5168 112-5169
Nitrile gloves, Microflex® 93-850
Gants
These nitrile disposable gloves are the ultimate barrier against harmful exposures and rip-tear. The tough, durable gloves stand up to tough jobs while significantly reducing workers’ risk of exposure to hazardous materials.
  • Two times more chemical splash protection due to breakthrough polymer bonding technique, when comparing EN 374 and ASTM F739 chemical permeation test data
  • Exceeds every known standard for barrier quality and consistency (Low 0,40 AQL)
  • Silicone-free formulation for product protection
  • Made with TNT™ Chemical Splash Resistance Technology, for soft durable protection against a wide range of chemicals
Ideal for blending and compounding materials, cleaning, filtration processes, laboratory analysis, light assembly tasks, paint and spray shops, pharmaceutical manufacturing, transfering liquids and solids.
Certifications: EN ISO 374-1:2016 Type B (JKOPT), EN ISO 374-5:2016 VIRUS; EN 421; Cat. III; AQL 0,40
Food contact approved
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité