10170.FI
10170.FIEA
1290
EUR
InStock
10170.FI
Erbium ≥99.9% (REO, base d'oxyde de terre rare), en feuilles, Thickness: 0.1 mm (0.004 in)
Erbium
Purity: 99.9%
CAS No.: 7440-52-0
Physical Form: ≈0.57g/25x25mm
Appearance: Silver-grey
UN No.: 3178
MDL No.: MFCD00010987