Dual-wire metal wristband, MagSnap 360™
Fournisseur: DESCO
19887
115-3036EA 39.3 EUR InStock
115-3036
Dual-wire metal wristband, MagSnap 360™
Dispositifs de protection antistatique Bracelets antistatiques
MagSnap 360™ dual-wire metal wristband creates a superior electrical connection to eliminate intermittent failures.
  • Magnetic connection between wristband and coil cord
  • Medical grade series stainless steel backplate and links
  • Date coded
Magnetic connection between wristband and coil cord creates a superior electrical connection, and eliminates intermittent failures in traditional snap connections using springs, extends the lifespan of the wrist strap, and maintains a 1 to 5 pound perpendicular breakaway force to prevent accidental disconnects. Wristband creates a redundant path-to-ground for an operator with resistance <3,5×10⁷ ohms per the required limits of ANSI/ESD S20.20 for Method 1. If one conductor is damaged, the operator will maintain the path-to-ground through the other conductor. Ideal for use with low voltage style monitors. Date coded so the lifespan of the product may be tracked to ensure quality control.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité