Gas detection tube for chloroprene
Fournisseur: Dräger
6718901
151-0138EA 125 EUR InStock
151-0138
Gas detection tube for chloroprene
Tubes de détection de gaz
Desorption stroke in clean air.
  • Data representative for use with an Accuro® hand operated bellows pump
  • Cross sensitivity: Many other organic compounds with C=C double bonds are indicated as well, but with different sensitivities
  • Impossible to measure chloroprene in the presence of dialkyl sulfides
One stroke is equivalent to a volume of 100 ±5 ml of gas passing through the tube.
Operating conditions: 10 to 30 °C ; 3 to 15 mg/l H₂O absolute humidity.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité