VWR®, Total Hardness Indicator, Tablets for Hardness of Water Determinations
Fournisseur: VWR Chemicals
160232EEA 49.6 EUR InStock
160232E
VWR®, Total Hardness Indicator, Tablets for Hardness of Water Determinations
Kits de tests environnementaux
Indicator tablets for determination of water hardness by titration with EDTA.
The tablets are dissolved in 100 ml of the water sample to be tested, which has been buffered at pH 10 with 1 ml of Ammonia/ Ammonium chloride buffer solution, and then titrated with EDTA solution (0,01 M) till the sample turns from red to blue.
Total hardness in mg/l = 10×ml of titrant used.
Control samples of calcium carbonate should be used to calibrate the method.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité