38878-50MG
38878-50MGEA
85.1
EUR
InStock
38878-50MG
Supelco® TraceCERT® Organic Reference Standard, Triadimenol
Étalons
Étalons organiques
Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
Attention: P202 - P260 - P263 - P273 - P202 - P260 - P263 - P273 - P301 + P312 - P308 + P313 - P301 + P312 - P308 + P313