Synonyms:
Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol
414719-4L
414719-4X4L
414719-4LEA
211
EUR
InStock
414719-4L
414719-4X4L
Méthanol ≥99.9%, CHROMASOLV™ pour analyse en GC capillaire, Riedel-de Haën™
Méthanol
- For GC headspace applications
- Meets USP, Ph. Eur. and ICH requirements
- GC headspace application tested
- Packed under inert gas for longer shelf life
Beilstein Number: 1098229