43455.06
43455.06EA
207
EUR
InStock
43455.06
Aluminium antimonide ≥99.99% (base métaux)
Aluminium antimonide
Purity: 99.99%
CAS No.: 25152-52-7
Molecular Formula: AlSb
Formula Weight: 148.73
Physical Form: 6mm & Down Pieces
Melting Point: 1050°
UN No.: 1549
MDL No.: MFCD00014154
In semiconductor technology