45605.09
45605.09EA
246
EUR
InStock
45605.09
Copper germanium selenide ≥99.99% (base métaux)
Copper germanium selenide
Purity: 99.99%
CAS No.: 54723-90-9
Molecular Formula: CuGeSe
Formula Weight: 215.12
Physical Form: 3-6mm Pieces
UN No.: UN3283
MDL No.: MFCD12546179