Synonyms:
Magnetite, Triiron tetraoxide, Iron(II,III) oxide
47141.14
47141.14EA
51.3
EUR
InStock
47141.14
Fer (II,III) oxyde 97%, nanopoudre
Fer (II,III) oxyde
Purity: 97%
CAS No.: 1317-61-9
Molecular Formula: Fe3O4
Formula Weight: 231.54
Physical Form: 50-100nm APS Powder, S. A. 20-50m2/g
Density: 5.2
Refractive Index: 3.0
Appearance: Black
Odour: Odourless
Melting Point: 1538°
MDL No.: MFCD00011010