50550-100MG
50550-100MGEA
107
EUR
InStock
50550-100MG
Supelco® TraceCERT® Organic Reference Standard, Pyrocatechol
Étalons
Étalons organiques
- GHS signal: GHS06, GHS08, GHS05
Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
Attention: P202 - P261 - P280 - P301 + P310 - P302 + P352 + P312 - P305 + P351 + P338