Membrane filters, cellulose acetate, Whatman™
Fournisseur: Whatman products (Cytiva)
7000-0002 7000-0004 7001-0004 10403012 10403112 10404001 10404006 10404012 10404014 10404026 10404031 10404044 10404106 10404112 10404114 10404126 10404131 10404180 10404139
514-0769EA 224 EUR InStock
514-0769 514-0770 514-0103 514-2322 514-0100 514-2350 514-2351 514-2352 514-2353 516-5015 514-2354 516-5016 514-2361 514-2362 514-2363 516-5018 514-2365 516-5020 516-5019
Membrane filters, cellulose acetate, Whatman™
Filtres Filtres à membrane
Cytiva's Whatman cellulose acetate membrane filters have very low protein binding, which minimizes sample loss when filtering protein-based aqueous samples.
  • Pure cellulose acetate composition, minimising sample contamination
  • Hydrophilic, for use with aqueous and alcoholic media
  • High solvent resistance, particularly to low molecular weight alcohols
  • High physical strength and a heat resistance up to 180 °C
  • Can be sterilized by most common methods without sacrificing membrane integrity
Due to their high chemical resistance, cellulose acetate membrane filters reliably prepare liquid scintillation cocktail for scintillation counter measurement.
High chemical resistance membranes.
Additionally, the highly heat resistant Whatman cellulose acetate membranes can filter hot liquids and hot gases.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité