VWR®, Screw Neck Vials ND18
Fournisseur: VWR Collection
548-3027AEA 667 EUR InStock
548-3027A 548-3277A 548-2484A
VWR®, Screw Neck Vials ND18
Fioles
Screw Neck ND18 vials manufactured from 1st hydrolytic class borosilicate glass with flat bottom. They can be purchased with a variety of polypropylene seal styles and septa materials.
  • High performance caps/septa
  • Good handling with gloves
  • Quick assembly
Screw Neck ND18 vials constructed from type I borosilicate glass with low metal content. Vials are available in clear with a flat bottom. Associated screw caps mostly available with assembled septa are engineered to prevent sample contamination while providing consistent, secure seals. The use of closed caps allows the samples to be stored for a long time (long-term storage applications).
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité