Supelco® Ascentis® Phenyl HPLC Columns
Fournisseur: Merck
581615-U 581616-U 581617-U 581618-U 581619-U 581620-U 581621-U
581615-UEA 481 EUR InStock
581615-U 581616-U 581617-U 581618-U 581619-U 581620-U 581621-U
Supelco® Ascentis® Phenyl HPLC Columns
Colonnes pour chromatographie
Ascentis®Phenyl HPLC columns are optimised to the three terms of the resolution equation: efficiency, retention, and selectivity.
  • High purity, type B silica for inertness, reproducibility, and stability
  • Modern bonding process optimises bonded phase coverage and maximises stability, while minimising bleed and unwanted secondary interactions
  • Compatible with LC-MS and all of today's sensitive instruments and methods
  • Scalable sensitivity from analytical to preparative
  • High surface area silica for high preparative loading capacity
  • Impurity: <5 ppm metals
  • Surface coverage: 5,2 µmol/m²
Ascentis® Phenyl phases are n-basic (electron donating) and are similar in overall retention to alkyl and EPG phases for easy column screening. The alternative selectivity of phenyl phases is often explained by the n-n interactions available through the phenyl ring. They provide low-bleed for MS or UV gradient applications due to the use of trifunctional bonding reagent, outstanding phenyl selectivity due to high phase loading and short butyl spacer and 100 % aqueous-compatible for highly-polar compounds.
Ascentis® bonded phases have a wide range of selectivities. It is likely that one or more Ascentis® phase will accomplish any small molecule HPLC separation. Ascentis® products cover all HPLC application areas, including the most sensitive trace-level analyses.
Information de livraison: Column test certificate available inside the box.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité