Single channel pipettes, electronic, variable volume, PIPETMAN® M Connected
Fournisseur: GILSON
F81044 F81045 F81046 F81043 F81040 F81047 F81041 F81042
613-6043EA 686 EUR InStock
613-6043 GILSF81045 613-6045 613-6042 613-6039 613-6046 613-6040 GILSF81042
Single channel pipettes, electronic, variable volume, PIPETMAN® M Connected
Pipettes Pipetteurs
PIPETMAN® M Connected is a Bluetooth®-enabled electronic pipette that offers accuracy and precision in both standard and repetitive pipetting modes. Requiring virtually no pipetting forces to aspirate and dispense samples, PIPETMAN M Connected helps reduce pipetting fatigue and increase pipetting efficiency. The pipette interacts in real-time with TRACKMAN® Connected making planning and executing pipetting protocols faster, easier, and more reliable.
  • Simple and intuitive motorized pipette
  • Ultimate consistency
  • Connect to Gilson Apps
With PipetteScope®, you’ll get access to service calibration, pipette activity, and useful resources for your pipette in one location. With PIPETMAN M Connected Software, create custom protocols from 11 different pipetting tasks and sync up to ten custom protocols into your pipette. TRACKMAN Connected check PIPETMAN M Connected calibration status and guides you through the execution of your protocol by displaying where, when, and how much to pipette with real-time interactions and automatic volume adjustments. At the end of the protocol, the application automatically generates a report detailing each of the steps performed with related pictures (e.g. for cells counts...) and notes taken during the execution.
Commander maintenant
COMMANDE
ACCESSOIRES
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité