pH meter, SevenDirect SD20
Fournisseur: Mettler - Toledo
30671550 30671554 30671555 30671556 30671557 30671558 30671559 30671560 30671561
662-2390EA 1120 EUR InStock
662-2390 662-2394 662-2395 662-2396 662-2397 662-2398 662-2399 662-2400 662-2402
pH meter, SevenDirect SD20
Compteurs électrochimiques pH-mètres et multi-paramètres
Robust bench top meter for accurate pH determinations in the lab and near production.
  • Large colour touch screen provides direct measurement and easy data entry
  • Instructions guide users through the sensor calibration process
  • Stores data, such as time stamp, sample and sensor ID with every measurement and prints or exports records automatically
  • Closed connector compartment and replaceable cover keep the instrument safe from dust and spills, even with sensors attached (IP 54 in-use)
The large colour touch screen offers a direct menu access, fast data entry and guides users through routine tasks. The instruments supports GLP compliance by storing time stamp, sample, sensor and method IDs with every measurement.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
ACCESSOIRES
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité