786-09
786-002
786-09T
786-424
786-264
786-165
786-09EA
284
EUR
InStock
786-09
786-002
786-09T
GENO786-424
GENO786-264
GENO786-165
picoLUCENT™ PLUS-HRP Kit
Réactifs d'électrophorèse
Western Blotting Reagents
The picoLUCENT™ PLUS-HRP kit is based on an ultra sensitive luminol substrate that produces chemiluminescence upon reaction with horseradish peroxidase. The chemiluminescence light emission can be recorded by a short exposure to autoradiography films.
- Low picogram detection (10⁻¹²) of antigens
- Low noise (signal/background ratio)
Information de livraison: Complete kit includes a non animal protein blocking agent (NAP-BLOCKER™) and wash buffer (femto-TBST™) and contains sufficient reagents for 25 mini blots (8×7,5 cm) or 1,500 cm² of PVDF or nitrocellulose membrane.