13420
AATB13420EA
230
EUR
InStock
AATB13420
Z-DEVD-AFC fluorogenic substrate
Z-DEVD-AFC
Z-DEVD-AFC is a fluorogenic substrate for caspase 3, a protease that is rapidly activated when cells are exposed to apoptotic conditions and that cleaves poly(ADP-ribose) polymerase.
- High fluorescence
- Rapid activation