1444
AATB1444EA
464
EUR
InStock
AATB1444
Tb maleimide, trFluor™
Tb maleimide
trFluor™ Tb probes are insensitive to fluorescence quenching when conjugated to biological polymers such as antibodies.
- Stable for 12 months
- Designed to enable time-resolved fluorometry (TRF)
Many biological compounds present in cells, serum or other biological fluids are naturally fluorescent, and thus the use of conventional, prompt fluorophores leads to serious limitations in assay sensitivity due to the high background caused by the autofluorescence of the biological molecules to be assayed.
Attention: For Research Use Only