Synonyms:
2-(3,6-Diacetoxy-9H-xanthen-9-yl)benzoic acid
15203
AATB15203EA
120
EUR
InStock
AATB15203
Dihydrofluorescein diacetate ≥95%
Dihydrofluorescein diacetate
Dihydrofluorescein diacetate (also called fluorescin diacetate) is hydrolyzed by cellular esterases to dihydrofluorescein (also called fluorescin) and is then oxidized to fluorescein primarily by H₂O₂.