16302
AATB16302EA
460
EUR
InStock
AATB16302
DAX-J2™ IR
DAX-J2™
DAX-J2™ IR is a new nitric oxide (NO) sensor recently developed by AAT Bioquest.
- Does not require esterase activity for no detection
- Ph-independent fluorescence
- More photostable than daf-2 product
- More sensitive for detecting no than daf-2
- Can be used for in vivo imaging of no
Attention: For research use only.