Synonyms:
POPO®-1, DiPO-1, NucPO-1 (434/456), POPO-1
17582
AATB17582EA
281
EUR
InStock
AATB17582
DiPO™-1 1 mM coloration d'acide nucléique
DiPO™-1
DiPO™-1 is the same compound to POPO®-1 (POPO® is the trademark of ThermoFisher).
- High affinity
- Optimized for flow cytometry and fluorescence microscopy