Synonyms:
Bis(N-methylacridinium) nitrate, N,N′-Dimethyl-9,9′-biacridinium dinitrate, 10,10'-Dimethyl-9,9'-biacridinium dinitrate
21259
AATB21259EA
120
EUR
InStock
AATB21259
Lucigenin chloride indicator
Lucigenin
A chloride indicator that has the fluorescence dependent of chloride concentration.
- Chloride-dependent fluorescence
- Superoxide detection
- Well-cited