25303
AATB25303EA
769
EUR
InStock
AATB25303
Cy3DIGE NHS ester ≥90%
Cy3DIGE NHS ester
Cy3DIGE NHS ester, an equivalent of Cy3® NHS ester minimal dye, is one of the major colors used for labeling proteins subject to DIGE analysis.
- Sensitive Detection
- Bright Fluorescence
Attention: For research use only.