3100
AATB3100EA
307
EUR
InStock
AATB3100
Cy5 biotin conjugate biodetection
Cy5 biotin conjugate
This bifunctional Cy5 biotin conjugate demonstrates excellent avidin binding and strong fluorescence.
- High fluorescence enhancement
- Sensitive detection
- Multiple applications