6910
AATB6910EA
292
EUR
InStock
AATB6910
CDPI3 Alkyne DNA binding
CDPI3 Alkyne
Minor Groove Binder (MGB) molecules selectively bind to the minor groove of a DNA molecule, the shallow furrow in the DNA helix.
- Stable binding
- Hybridization probe precursor