AB285231-96
ABCAAB285231-96EA
800
EUR
InStock
ABCAAB285231-96
Creatine Kinase MB (CK-MB) (Mouse) ELISA Kit
Tests
Tests ELISA
A Sandwich ELISA kit for the quantitative measurement of creatine kinase MB in biological fluids.
- Detection range: 78,125 to 5000 pg/ml
- Sensitivity: <46,875 pg/ml
- Assay precision: Intra-assay: CV <8%; Inter-assay: CV <10%
- No significant cross-reactivity or interference was observed
Easy, convenient and time-saving method to measure the level of Creatine Kinase MB in serum, plasma, tissue homogenates, culture supernatants and other biological fluids.
Creatine kinase (CK) also known as creatine phosphokinase (CPK) and ATP: creatine N-phosphotransferase is a common cellular enzyme. It catalySes the reversible conversion of creatine and ATP into ADP and phosphocreatine. CK is widely expressed in various tissues and cell types, with highest activity in striated muscles, heart tissue and brain.
CK consists of two subunits: M (muscle) and B (brain), and has three isoenzymes: CK-MM (skeleton muscle), CK-MB (cardiac muscle), and CK-BB (brain). Increased CK level is associated with many diseases such as myocardial infarction, muscular dystrophy, pulmonary infarction and brain tumors.
Accurate measurement of CK is crucial for early diagnosis, prediction and therapeutic strategy. CK-MB ELISA kit is a sandwich ELISA assay for the quantitative measurement of CK-MB in mouse serum, plasma and cell culture supernatants. The density of colour is proportional to the amount of CK-MB captured from the samples.