AB285277-96
ABCAAB285277-96EA
990
EUR
InStock
ABCAAB285277-96
Cathepsin G (CTSG) (Human) ELISA Kit
Tests
Tests ELISA
A sandwich ELISA kit for the quantitative measurement of cathepsin G in biological fluids.
- Easy, convenient and time-saving method
- Detection Range: 0,156 to 10 ng/ml
- Sensitivity: <0,094 pg/ml
- Assay precision: Intra-assay: CV <8%; inter-assay: CV <10%
- No significant cross-reactivity or interference was observed
- 100 Assays per kit
Cathepsin G (CTSG) is an enzymatic protein belonging to the peptidase or protease families. The protein is found in azurophil granules of neutrophilic polymorphonuclear leukocytes.
The encoded protease has a specificity similar to that of Chymotrypsin C, and may participate in the killing and digestion of engulfed pathogens, and in connective tissue remodeling at sites of inflammation.
CTSG ELISA kit is a sandwich ELISA assay for the quantitative measurement of human CTSG in serum, plasma, and cell culture supernatants.
The density of color is proportional to the amount of human CTSG captured from the samples.