AB285297-96
ABCAAB285297-96EA
835
EUR
InStock
ABCAAB285297-96
IL-11 (Human) ELISA Kit
Tests
Tests ELISA
An Sandwich ELISA kit for quantifying human IL-11 in human serum, plasma & other biological fluids.
- Easy, convenient and time-saving method to measure the level of IL-11 in human serum, plasma, tissue homogenates, culture supernatants and other biological fluids
- Detection Range: 31,25 to 2000 pg/ml
- Sensitivity: <18,75 pg/ml
- Assay Precision: Intra-Assay: CV <8%; Inter-Assay: CV <10%
IL-11 is a cytokine that stimulates the proliferation of hematopoietic stem cells and megakaryocyte progenitor cells and induces megakaryocyte maturation resulting in increased platelet production. Also promotes the proliferation of hepatocytes in response to liver damage. Binding to its receptor formed by IL6ST and either IL11RA1 or IL11RA2 activates a signaling cascade that promotes cell proliferation. Signaling leads to the activation of intracellular protein kinases and the phosphorylation of STAT3. BioVision’s IL-11 ELISA kit is a sandwich ELISA assay for the quantitative measurement of human IL-9 in serum, plasma and cell culture supernatants. The density of color is proportional to the amount of human IL-11 captured from the samples.