QuickDetect™ 8-OHdG (Rat) ELISA Kit
Fournisseur: Abcam
AB285302-96
ABCAAB285302-96EA 710 EUR InStock
ABCAAB285302-96
QuickDetect™ 8-OHdG (Rat) ELISA Kit
Tests Tests ELISA
A rapid sandwich ELISA kit for quantify 8-OHdG in rat serum, plasma, urine and other biological fluid.
  • Easy, convenient and time-saving method to measure the level of 8-OHdG in rat serum, plasma, urine, cell culture samples, tissue samples
  • Detection Range: 22 to 1800 pg/ml
  • Sensitivity: <5 pg/ml
8-hydroxy-2'-deoxyguanosine (8-OHdG) is an oxidised derivative of deoxyguanosine. Concentrations of 8-oxo-dG within a cell are a measurement of oxidative stress. It is considered as a critical biomarker of oxidative stress and carcinogenesis. This ELISA kit uses Competitive-ELISA as the method. The microtiter plate provided in this kit has been pre-coated with 8-OHdG. During the reaction,8-OHdG in the sample or standard competes with a fixed amount of 8-OHdG on the solid phase supporter for sites on the Biotinylated Detection Antibody specific to 8-OHdG. BioVision’s 8-OHdG ELISA kit is a sandwich ELISA assay for the quantitative measurement of Rat 8-OHdG in serum, plasma and cell culture supernatants in 90 minutes. The density of colour is proportional to the amount of Rat 8-OHdG captured from the samples.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité