KA2021
ABNOKA2021EA
791
EUR
InStock
ABNOKA2021
B2m (Rat) ELISA kit
Tests
Tests ELISA
B2m (Rat) ELISA Kit is an enzyme immunoassay used for the quantitative determination of Beta 2-Microglobulin (B2m) in rat biological fluids.
- Calibration range: 0,156 to 5 ng/ml
- Sample volume: 100 µl
Information de livraison: Kit contains Anti-Rat B2M ELISA 96-well microplate (8×12), calibrator (1 vial), running buffer (5×50 ml), washing solution concentrate (20×50 ml), enzyme-antibody conjugate (100×150 µl), chromogen-substrate solution (12 ml) and stop solution (12 ml).