IL8 (Human) ELISA Kit
Fournisseur: Abnova
KA3068
ABNOKA3068EA 791 EUR InStock
ABNOKA3068
IL8 (Human) ELISA Kit
Tests Tests ELISA
IL8 (Human) ELISA Kit is intended for the quantitative measurement of human IL8.
  • Suitable sample - Body fluid, cell culture supernatant, plasma, serum, tissue lysate
  • Assay type - Quantitative
  • Calibration range - 15,6 to 1000 pg/ml (Body fluid, tissue lysate, cell culture supernate); 7,8 to 500 pg/ml (Serum, plasma)
Quality control testing:
Standard curve - The standard curve is for the purpose of illustration only and should not be used to calculate unknowns. A standard curve should be generated each time the assay is performed.
The protein encoded by this gene is a member of the CXC chemokine family. This chemokine is one of the major mediators of the inflammatory response. This chemokine is secreted by several cell types. It functions as a chemoattractant, and is also a potent angiogenic factor. This gene is believed to play a role in the pathogenesis of bronchiolitis, a common respiratory tract disease caused by viral infection. This gene and other ten members of the CXC chemokine gene family form a chemokine gene cluster in a region mapped to chromosome 4q.
T cell chemotactic factor|beta-thromboglobulin-like protein|chemokine (C-X-C motif) ligand 8|emoctakin|granulocyte chemotactic protein 1|lymphocyte-derived neutrophil-activating factor|monocyte-derived neutrophil chemotactic factor|neutrophil-activating p.
Attention: For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité