KA6696.1KIT
ABNOKA6696.1KITEA
359
EUR
InStock
ABNOKA6696.1KIT
Helicobacter pylori IgG ELISA kit
Tests
Tests ELISA
Helicobacter pylori IgG ELISA Kit is an enzyme immunoassay for the quantitative measurement of IgG antibodies to Helicobacter pylori in human serum and plasma.