KA6698.1KIT
ABNOKA6698.1KITEA
359
EUR
InStock
ABNOKA6698.1KIT
Cytomegalovirus IgG ELISA kit
Tests
Tests ELISA
Cytomegalovirus IgG ELISA Kit is a sandwich enzyme immunoassay for the quantitative and qualitative measurement of Cytomegalovirus IgG in human serum and plasma.