KA6707.1KIT
ABNOKA6707.1KITEA
359
EUR
InStock
ABNOKA6707.1KIT
Cytomegalovirus IgM ELISA kit
Tests
Tests ELISA
Cytomegalovirus IgM ELISA Kit is an enzyme immunoassay for the quantitative and qualitative measuremnet of IgM antibodies to Cytomegalovirus (CMV) in human serum and plasma.