KA6728.1KIT
ABNOKA6728.1KITEA
359
EUR
InStock
ABNOKA6728.1KIT
Epstein-Barr Virus EBNA-1 IgG ELISA kit
Tests
Tests ELISA
Epstein-Barr Virus EBNA-1 IgG ELISA Kit is a qualitative determination of IgG class antibodies against Epstein-Barr virus nuclear antigen type 1 (EBV-EBNA-1) in human serum or plasma.