KA6779.1KIT
ABNOKA6779.1KITEA
359
EUR
InStock
ABNOKA6779.1KIT
Influenza Virus A IgG (Human) ELISA kit
Tests
Tests ELISA
Influenza Virus A IgG (Human) ELISA Kit is a qualitative determination of IgG class antibodies against Influenza Virus A in human serum or plasma.