KA6781.1KIT
ABNOKA6781.1KITEA
359
EUR
InStock
ABNOKA6781.1KIT
Measles Virus IgG (Human) ELISA kit
Tests
Tests ELISA
Measles Virus IgG (Human) ELISA Kit is a qualitative determination of IgG class antibodies against Measles Virus in human serum or plasma.