KA6788.1KIT
ABNOKA6788.1KITEA
791
EUR
InStock
ABNOKA6788.1KIT
Ovalbumin Specific IgE (Human) ELISA kit
Tests
Tests ELISA
Ovalbumin Specific IgE (Human) ELISA Kit is for the qualitative determination of human ovalbumin specific IgE concentrations in serum or plasma.