Recombinat Protein,Human Apolipoprotein E APOE Protein, His Tag,Human Apolipoprotein E, His Tag (APE-H5246) is expressed from human 293 cells (HEK293). It contains AA Lys 19 - His 317 (Accession P02649-1).This protein carries a polyhistidine tag at s 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human Apolipoprotein E APOE Protein, His Tag,Human Apolipoprotein E, His Tag (APE-H5246) is expressed from human 293 cells (HEK293). It contains AA Lys 19 - His 317 (Accession P02649-1).This protein carries a polyhistidine tag at s 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human Apolipoprotein E APOE Protein, His Tag,Human Apolipoprotein E, His Tag (APE-H5246) is expressed from human 293 cells (HEK293). It contains AA Lys 19 - His 317 (Accession P02649-1).This protein carries a polyhistidine tag at s 1 * 100 µG ACRBAPE-H5246100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou