Cynomolgus Rhesus macaque B7-1, His Tag (HPLC-verified) (CD0-C52H3) is expressed from human 293 cells (HEK293). It contains AA Val 35 - Asn 242 (Accession G7NXN7). In the region Val 35 - Asn 242, the AA sequence of Cynomolgus and Rhesus macaque B7-1 1 * 100 µG
Fournisseur: ACROBIOSYSTEMS INC
COMMANDE
Développer tout / Réduire tout
Cynomolgus Rhesus macaque B7-1, His Tag (HPLC-verified) (CD0-C52H3) is expressed from human 293 cells (HEK293). It contains AA Val 35 - Asn 242 (Accession G7NXN7). In the region Val 35 - Asn 242, the AA sequence of Cynomolgus and Rhesus macaque B7-1 1 * 100 µG ACRBCD0-C52H3100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou