Human Transferrin R, His Tag (CD1-H5243) is expressed from human 293 cells (HEK293). It contains AA Cys 89 - Phe 760 (Accession P02 786-1). 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Human Transferrin R, His Tag (CD1-H5243) is expressed from human 293 cells (HEK293). It contains AA Cys 89 - Phe 760 (Accession P02 786-1). 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Human Transferrin R, His Tag (CD1-H5243) is expressed from human 293 cells (HEK293). It contains AA Cys 89 - Phe 760 (Accession P02 786-1). 1 * 1 mg ACRBCD1-H5243-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou