Human CD27 Ligand, Fc Tag (active trimer) (MALS verified) (CDL-H5266) is expressed from human 293 cells (HEK293). It contains AA Ser 52 - Pro 193 (Accession P32970-1) trimer Design. 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Human CD27 Ligand, Fc Tag (active trimer) (MALS verified) (CDL-H5266) is expressed from human 293 cells (HEK293). It contains AA Ser 52 - Pro 193 (Accession P32970-1) trimer Design. 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Human CD27 Ligand, Fc Tag (active trimer) (MALS verified) (CDL-H5266) is expressed from human 293 cells (HEK293). It contains AA Ser 52 - Pro 193 (Accession P32970-1) trimer Design. 1 * 100 µG ACRBCDL-H5266100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou